Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triphenylmethyl mercaptan, 98+%

Click to view available options
Quantity:
5g
25g
100g
3695-77-0
C19H16S
276.397
MFCD00004854
JQZIKLPHXXBMCA-UHFFFAOYSA-N
triphenylmethyl mercaptan,trityl mercaptan,tritylthiol,triphenylmethylmercaptan,methanethiol, triphenyl,tritylmercaptan,triphenyl-methanethiol,alpha,alpha-diphenylbenzenemethanethiol,benzenemethanethiol, alpha,alpha-diphenyl,benzenemethanethiol, .alpha.,.alpha.-diphenyl
77281
triphenylmethanethiol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
3695-77-0
C19H16S
276.397
MFCD00004854
JQZIKLPHXXBMCA-UHFFFAOYSA-N
triphenylmethyl mercaptan,trityl mercaptan,tritylthiol,triphenylmethylmercaptan,methanethiol, triphenyl,tritylmercaptan,triphenyl-methanethiol,alpha,alpha-diphenylbenzenemethanethiol,benzenemethanethiol, alpha,alpha-diphenyl,benzenemethanethiol, .alpha.,.alpha.-diphenyl
77281
triphenylmethanethiol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
Applications
Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Quantity 5g
Melting Point 103°C to 108°C
Odor Stench
Beilstein 2113168
Solubility Information It is insoluble in water.
Formula Weight 276.4
Percent Purity ≥98%
Chemical Name or Material Triphenylmethyl mercaptan

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P271-P304+P340-P312

H332

EINECSNumber : 223-020-3

RTECSNumber : PB5335000

TSCA : Yes

Recommended Storage : Ambient temperatures