Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylmethylebromide, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
596-43-0
C19H15Br
323.22
MFCD00000120
NZHXEWZGTQSYJM-UHFFFAOYSA-N
bromotriphenylmethane,triphenylmethyl bromide,trityl bromide,triphenylbromomethane,bromodiphenylmethyl benzene,methane, bromotriphenyl,benzene, 1,1',1-bromomethylidyne tris,triphenylmethylbromide,bromomethanetriyl tribenzene,2-bromo-1,1,1-triphenylethane
11692
[bromo(diphenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br
596-43-0
C19H15Br
323.22
MFCD00000120
NZHXEWZGTQSYJM-UHFFFAOYSA-N
bromotriphenylmethane,triphenylmethyl bromide,trityl bromide,triphenylbromomethane,bromodiphenylmethyl benzene,methane, bromotriphenyl,benzene, 1,1',1-bromomethylidyne tris,triphenylmethylbromide,bromomethanetriyl tribenzene,2-bromo-1,1,1-triphenylethane
11692
[bromo(diphenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br
Boiling Point 230°C (15.0mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 10g
Melting Point 151°C to 155°C
Assay Percent Range 98%
Linear Formula (C6H5)3CBr
Beilstein 05, 704
Fieser 01,1254; 16,375
Solubility Information Solubility in water: insoluble
Formula Weight 323.22
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Triphenylmethyl bromide
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger