Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
603-35-0
C18H15P
262.28
MFCD00003043
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
triphenylphosphine,triphenyl phosphine,phosphine, triphenyl,triphenylphosphorus,triphenyl-phosphane,triphenylphosphide,phosphorustriphenyl,trifenylfosfin,trifenylfosfin czech,triphenylphosphine resin
11776
triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
603-35-0
C18H15P
262.28
MFCD00003043
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
triphenylphosphine,triphenyl phosphine,phosphine, triphenyl,triphenylphosphorus,triphenyl-phosphane,triphenylphosphide,phosphorustriphenyl,trifenylfosfin,trifenylfosfin czech,triphenylphosphine resin
11776
triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
Color White
Infrared Spectrum Authentic
Flash Point 181°C
Quantity 1kg
Melting Point 78.5°C to 81.5°C
Boiling Point 377.0°C
Packaging Plastic bottle
Assay Percent Range 99%
Linear Formula (C6H5)3P
Beilstein 16,759
Fieser 01,1238; 02,443; 03,317; 04,548; 05,725; 06,643; 07,403; 11,588; 12,550; 13,308; 14,60; 15,248; 16,264; 17,389
Merck Index 15,9916
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in glacial acetic acid,, soluble in benzene, chloroform, acetone and ccl4,, freely soluble in diethyl ether,, slightly soluble in alcohol
Formula Weight 262.28
Physical Form Crystals, Crystalline Powder or Flakes
Percent Purity 99%
Chemical Name or Material Triphenylphosphine
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-036-0

RTECSNumber : SZ3500000

TSCA : TSCA