Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine Borane 97.0+%, TCI America™

Click to view available options
Quantity:
25g
2049-55-0
C18H15BP
273.102
MFCD00012427
QFLQJPFCNMSTJZ-UHFFFAOYSA-N
triphenylphosphine borane,borane triphenylphosphine complex,triphenylphosphineborane,boron; triphenylphosphane,triphenylphosphinborane,pubchem18246,acmc-209fav,borane-triphenylphosphine complex,triphenylphosphine boranylidyne
6364612
boron;triphenylphosphane
[B].C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
2049-55-0
C18H15BP
273.102
MFCD00012427
QFLQJPFCNMSTJZ-UHFFFAOYSA-N
triphenylphosphine borane,borane triphenylphosphine complex,triphenylphosphineborane,boron; triphenylphosphane,triphenylphosphinborane,pubchem18246,acmc-209fav,borane-triphenylphosphine complex,triphenylphosphine boranylidyne
6364612
boron;triphenylphosphane
[B].C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
Quantity 25g
Color White-Yellow
Formula Weight 276.13
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material Triphenylphosphine Borane