Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triphenylphosphine diiodide, tech. 90%

Manufacturer:  Alfa Aesar™ H5558506

Quantity 5g

 View more versions of this product

Catalog No. AAH5558506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6396-07-2
516.1
UN2923
Diiodotriphenylphosphorane
Technical Grade
(C6H5)3PI2
MFCD00216568
2740417
Triphenylphosphine diiodide

Specifications

6396-07-2
(C6H5)3PI2
MFCD00216568
2740417
Yellow to orange
5g
Triphenylphosphine diiodide
Technical Grade
Powder
516.1
UN2923
Diiodotriphenylphosphorane
210° to 211°C
Light Sensitive
90%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : TB6100000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit