Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine oxide, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
791-28-6
C18H15OP
278.29
FIQMHBFVRAXMOP-UHFFFAOYSA-N
triphenylphosphine oxide,phosphine oxide, triphenyl,triphenyl phosphorus oxide,triphenyl phosphine oxide,triphenylphosphane oxide,tppo,triphenylphosphineoxide,ph3po,diphenylphosphoroso benzene,triphenylphosphanoxid
13097
CHEBI:36601
diphenylphosphorylbenzene
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC=CC=C3
791-28-6
C18H15OP
278.29
FIQMHBFVRAXMOP-UHFFFAOYSA-N
triphenylphosphine oxide,phosphine oxide, triphenyl,triphenyl phosphorus oxide,triphenyl phosphine oxide,triphenylphosphane oxide,tppo,triphenylphosphineoxide,ph3po,diphenylphosphoroso benzene,triphenylphosphanoxid
13097
CHEBI:36601
diphenylphosphorylbenzene
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC=CC=C3
Color Brown-Pink to White
Melting Point 154.0°C to 158.0°C
Infrared Spectrum Authentic
Boiling Point >360.0°C
Quantity 500g
Flash Point 180°C
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula (C6H5)3P(O)
Beilstein 16,IV,1011
Fieser 14,337; 16,368
Solubility Information Solubility in water: slightly soluble.
Formula Weight 278.29
Physical Form Crystalline Powder or Flakes
Percent Purity 99%
Chemical Name or Material Triphenylphosphine oxide
Show More Show Less