Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g, 100-200 mesh, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
39319-11-4
C18H15P
262.292
MFCD00148025
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
11776
triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
39319-11-4
C18H15P
262.292
MFCD00148025
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
11776
triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3
Packaging Glass bottle
Quantity 5g
Physical Form 100 To 200 Mesh
Chemical Name or Material Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g