Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triphenylphosphine sulfide, 97%

Manufacturer:  Alfa Aesar™ 10177-22

Quantity 100g

 View more versions of this product

Catalog No. AA1017722

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

3878-45-3
MFCD00004917
Triphenylphosphine sulfide
294.34
UN3464

Specifications

3878-45-3
294.34
UN3464
100g
97%
Crystalline
MFCD00004917
157° to 160°C
Triphenylphosphine sulfide

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P311-P362-P301+P312-P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit