Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphinecopper(I) hydride hexamer, 96%, Acros Organics™

Click to view available options
Quantity:
1g
5g
33636-93-0
C108H90Cu6P6
1955.027
MFCD00221518
KVBHFBAACGHMKE-UHFFFAOYSA-N
stryker's reagent,triphenylphosphine copper hydride hexamer,triphenylphosphine-copper i hydride hexamer,unii-8x3r32m7xu,hydrido triphenylphosphine copper i hexamer,stryker's reagent mi,triphenylphosphine cuprous hydride,triphenylphosphine copper i hydride hexamer,copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro,cuprous hydride triphenylphosphine hexamer
11982471
copper;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cu].[Cu].[Cu].[Cu].[
33636-93-0
C108H90Cu6P6
1955.027
MFCD00221518
KVBHFBAACGHMKE-UHFFFAOYSA-N
stryker's reagent,triphenylphosphine copper hydride hexamer,triphenylphosphine-copper i hydride hexamer,unii-8x3r32m7xu,hydrido triphenylphosphine copper i hexamer,stryker's reagent mi,triphenylphosphine cuprous hydride,triphenylphosphine copper i hydride hexamer,copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro,cuprous hydride triphenylphosphine hexamer
11982471
copper;triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cu].[Cu].[Cu].[Cu].[
Reduction reagent
Quantity 5g
Infrared Spectrum Authentic
Color Red
NMR 96.0% min.
Assay Percent Range 97%
Merck Index 15, 8987
Solubility Information Solubility in water: reacts slowly.
Formula Weight 1961.07
Physical Form Crystals
Percent Purity 96%
Chemical Name or Material Triphenylphosphinecopper(I) hydride hexamer
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning