Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triphenylphosphoranylidene)acetonitrile, 96%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
16640-68-9
C20H16NP
301.33
MFCD00567633
APISVOVOJVZIBA-UHFFFAOYSA-N
triphenylphosphoranylidene acetonitrile,2-triphenylphosphoranylidene acetonitrile,cyanomethylene triphenylphosphorane,acetonitrile, triphenylphosphoranylidene,2-triphenyl-$l^ 5-phosphanylidene acetonitrile,2-triphenyl-??-phosphanylidene acetonitrile,acmc-1c6hd,ksc493a4b,cyanomethylenetriphenylphosphorane
85524
2-(triphenyl-$l^{5}-phosphanylidene)acetonitrile
C1=CC=C(C=C1)P(=CC#N)(C2=CC=CC=C2)C3=CC=CC=C3
16640-68-9
C20H16NP
301.33
MFCD00567633
APISVOVOJVZIBA-UHFFFAOYSA-N
triphenylphosphoranylidene acetonitrile,2-triphenylphosphoranylidene acetonitrile,cyanomethylene triphenylphosphorane,acetonitrile, triphenylphosphoranylidene,2-triphenyl-$l^ 5-phosphanylidene acetonitrile,2-triphenyl-??-phosphanylidene acetonitrile,acmc-1c6hd,ksc493a4b,cyanomethylenetriphenylphosphorane
85524
2-(triphenyl-$l^{5}-phosphanylidene)acetonitrile
C1=CC=C(C=C1)P(=CC#N)(C2=CC=CC=C2)C3=CC=CC=C3
Packaging Glass bottle
Quantity 5g
Melting Point 189.0°C to 198.0°C
Assay Percent Range 95% min. (HPLC)
Linear Formula (C6H5)3P=CHCN
Formula Weight 301.33
Percent Purity 96%
Chemical Name or Material (Triphenylphosphoranylidene)acetonitrile

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

EINECSNumber : 240-689-7