Alfa Aesar™ Triphenylsulfonium hexafluoroarsenate, As 14.5% min

Manufacturer: Alfa Aesar™ 2084706

Quantity 5g

 View more versions of this product

Catalog No. AA2084706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57900-42-2
    Linear Formula (C6H5)3SAsF6
    Formula Weight 452.29
    MDL Number MFCD00061401
    UN Number UN1557
    Chemical Name or Material Triphenylsulfonium hexafluoroarsenate

    Specifications

    Formula Weight 452.29
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00061401
    CAS 57900-42-2
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Melting Point 195° to 197°C
    Quantity 5g
    Physical Form Powder
    Linear Formula (C6H5)3SAsF6
    UN Number UN1557
    Health Hazard 1 Danger
    Color White
    Sensitivity Moisture Sensitive
    TSCA TSCA
    Concentration As 14.5% min.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    Chemical Name or Material Triphenylsulfonium hexafluoroarsenate

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit