Alfa Aesar™ Triphenyltin acetate, 97%

Manufacturer: Alfa Aesar™ 45789-18

Quantity 50g

 View more versions of this product

Catalog No. AA4578918

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 900-95-8
    Molecular Formula C20H18O2Sn
    Linear Formula (C6H5)3SnOOCCH3
    Formula Weight 409.05
    MDL Number MFCD00014977
    UN Number UN3146
    Beilstein 4146107
    Merck Index 14,9745
    Synonym Fentin acetate; Triphenylacetoxytin
    Chemical Name or Material Triphenyltin acetate

    Specifications

    Formula Weight 409.05
    Physical Form Powder
    Molecular Formula C20H18O2Sn
    MDL Number MFCD00014977
    Quantity 50g
    Beilstein 4146107
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H372-H351-H361-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    S
    Color White
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    Linear Formula (C6H5)3SnOOCCH3
    CAS 900-95-8
    UN Number UN3146
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and
    RTECS Number WH6650000
    Melting Point 121° to 123°C
    Merck Index 14,9745
    Synonym Fentin acetate; Triphenylacetoxytin
    Chemical Name or Material Triphenyltin acetate
    Assay 97%

    GHS H Statement
    H301-H311-H330-H372-H351-H361-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    S

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit