Alfa Aesar™ Triphenyltin hydroxide

Manufacturer: Alfa Aesar™ 7117722

Quantity 100g

 View more versions of this product

Catalog No. AA7117722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76-87-9
    Molecular Formula C18H16OSn
    Linear Formula (C6H5)3SnOH
    Formula Weight 367.01
    MDL Number MFCD00013928
    UN Number UN3146
    Merck Index 14,9745
    Synonym Hydroxytriphenyltin
    Chemical Name or Material Triphenyltin hydroxide

    Specifications

    Formula Weight 367.01
    Health Hazard 2 GHS H Statement
    H301-H311-H330-H372-H351-H361-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 76-87-9
    Physical Form Powder
    Molecular Formula C18H16OSn
    MDL Number MFCD00013928
    Quantity 100g
    UN Number UN3146
    Health Hazard 1 Danger
    Color Off-white
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    Melting Point 122° to 124°C
    Linear Formula (C6H5)3SnOH
    Merck Index 14,9745
    Synonym Hydroxytriphenyltin
    Solubility Information Slightly soluble in alcohol, toluene
    RTECS Number WH8575000
    Chemical Name or Material Triphenyltin hydroxide

    GHS H Statement
    H301-H311-H330-H372-H351-H361-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit