Alfa Aesar™ Tripotassium citrate monohydrate, 97%

Manufacturer: Alfa Aesar™ 4503836

Quantity 500g

 View more versions of this product

Catalog No. AA4503836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6100-05-6
    Molecular Formula K3C6H5O7·H2O
    Formula Weight 324.42 (306.41 Anhydrous)
    MDL Number MFCD00150442
    Merck Index 14,7623
    Synonym Citric acid tripotassium salt monohydrate; Potassium citrate tribasic monohydrate
    Chemical Name or Material Tripotassium citrate monohydrate

    Specifications

    Physical Form Lump/Granular
    Formula Weight 324.42 (306.41 Anhydrous)
    MDL Number MFCD00150442
    Synonym Citric acid tripotassium salt monohydrate; Potassium citrate tribasic monohydrate
    TSCA TSCA
    Molecular Formula K3C6H5O7·H2O
    CAS 6100-05-6
    Merck Index 14,7623
    Melting Point 230°C (dec.)
    Density 1.98g/mL
    Quantity 500g
    Chemical Name or Material Tripotassium citrate monohydrate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit