Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TrippNT Acrylic Glove Box Holders Non-distribution product as customer accommodation.

$63.60 - $117.60

Specifications

Color Clear
Material Acrylic
For Use With (Application) Ideal for labs, clinics, exam rooms, and other industrial settings
Includes Keyhole or magnet mount
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
NC9990259 Non-distribution product as customer accommodation.
View Documents
TrippNT
50850
Quadruple Each for $117.60
Add to cart
 
NC0263498 Non-distribution product as customer accommodation.
View Documents
TrippNT
50831
Single Each for $63.60
Add to cart
 
Specifications

Specifications

Clear
Acrylic
Ideal for labs, clinics, exam rooms, and other industrial settings
Keyhole or magnet mount
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit