Learn More
TrippNT Portable Personal Desiccator
TrippNT 50140 Portable Personal Desiccator is designed to store, carry and keep dry samples and chemicals in an acrylic, see-through container. Removable perforated tray lifts out easily. 16x5x6 inches (width height depth) 4 pounds
Manufacturer:  TrippNT 50140
Encompass
Catalog No. NC9779153
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TrippNT 50140 Portable Personal Desiccator is designed to store, carry and keep dry samples and chemicals in an acrylic, see-through container. Removable perforated tray lifts out easily. 16x5x6 inches (width height depth) 4 pounds
Specifications
Specifications
Portable Personal Desiccator
6 in.
5 in.
Removable perforated tray
Store, carry and keep dry samples and chemicals
16 in.