Learn More
TrippNT Radioactive Waste Shield
Manufacturer: TrippNT 50440
Description
Designed for maximum protection against beta-emitting isotopes.
Specifications
4 industrial casters; 2 with brakes | |
Protects against Beta emitters Tritium, C-14 and P-32 | |
15 in. | |
24 in. | |
0.5 in. | |
13.25 in. |
Mobile Hazardous Waste Container | |
Acrylic | |
13.25 in. | |
19.25 in. | |
15 in. | |
Standard waste boxes |