Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triprolidine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
6138-79-0
C19H25ClN2O
332.87
CUZMOIXUFHOLLN-UMVVUDSKSA-N
hydrogen 2-[(1E)-1-(4-methylphenyl)-3-(pyrrolidin-1-yl)prop-1-en-1-yl]pyridine hydrate chloride
[H+].O.[Cl-].CC1=CC=C(C=C1)C(=C/CN1CCCC1)\C1=CC=CC=N1
6138-79-0
C19H25ClN2O
332.87
CUZMOIXUFHOLLN-UMVVUDSKSA-N
hydrogen 2-[(1E)-1-(4-methylphenyl)-3-(pyrrolidin-1-yl)prop-1-en-1-yl]pyridine hydrate chloride
[H+].O.[Cl-].CC1=CC=C(C=C1)C(=C/CN1CCCC1)\C1=CC=CC=N1

T1127, 6138-79-0, C19H22N2•HCl•H2O

Spectrum™ Triprolidine Hydrochloride, USP is an antihistamine used to relieve symptoms of allergy, hay fever, and the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 25 g
Ignition Residue 0.001
Water 4-6%
CAS Max % 1
Percent Purity 98 to 102%
Grade USP