Alfa Aesar™ Tripropargylamine, 97%

Manufacturer: Alfa Aesar™ H5345914

Quantity 25g

 View more versions of this product

Catalog No. AAH5345914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6921-29-5
    Molecular Formula C9H9N
    Linear Formula (HC≡CCH2)3N
    Formula Weight 131.18
    MDL Number MFCD00008577
    UN Number UN1993
    Chemical Name or Material Tripropargylamine

    Specifications

    Formula Weight 131.18
    Linear Formula (HC≡CCH2)3N
    Boiling Point 79° to 85°C (11mmHg)
    Quantity 25g
    Molecular Formula C9H9N
    MDL Number MFCD00008577
    CAS 6921-29-5
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 57°C (135°F)
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    RTECS Number YK7660000
    Chemical Name or Material Tripropargylamine
    Assay 97%

    Description

    Applications
    Tripropargylamine is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit