Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropyl orthoformate, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
621-76-1
C10H22O3
190.29
MFCD00015214
RWNXXQFJBALKAX-UHFFFAOYSA-N
tripropyl orthoformate,tri-n-propyl orthoformate,orthoformic acid tripropyl ester,1-dipropoxymethoxy propane,unii-z8f0df6af9,propane, 1,1',1-methylidynetris oxy tris,z8f0df6af9,tripropoxymethane,tripropylorthoformate
69311
1-(dipropoxymethoxy)propane
CCCOC(OCCC)OCCC
621-76-1
C10H22O3
190.29
MFCD00015214
RWNXXQFJBALKAX-UHFFFAOYSA-N
tripropyl orthoformate,tri-n-propyl orthoformate,orthoformic acid tripropyl ester,1-dipropoxymethoxy propane,unii-z8f0df6af9,propane, 1,1',1-methylidynetris oxy tris,z8f0df6af9,tripropoxymethane,tripropylorthoformate
69311
1-(dipropoxymethoxy)propane
CCCOC(OCCC)OCCC
Alkylation reagent
Boiling Point 106°C to 108°C (40.0mmHg)
Flash Point 72°C
Specific Gravity 0.879
Refractive Index 1.4060 to 1.408
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
Quantity 25mL
Density 0.8790g/mL
Assay Percent Range 98%
Linear Formula CH(OCH2CH2CH3)3
Beilstein 02, IV, 27
Formula Weight 190.29
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Tripropyl orthoformate
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.