Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, ≥97%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
510758-28-8
C30H30N10
530.64
MFCD09265124
WKGZJBVXZWCZQC-UHFFFAOYSA-N
tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine
11203363
1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
C1=CC=C(C=C1)CN2C=C(N=N2)CN(CC3=CN(N=N3)CC4=CC=CC=C4)CC5=CN(N=N5)CC6=CC=CC=C6
510758-28-8
C30H30N10
530.64
MFCD09265124
WKGZJBVXZWCZQC-UHFFFAOYSA-N
tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine
11203363
1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
C1=CC=C(C=C1)CN2C=C(N=N2)CN(CC3=CN(N=N3)CC4=CC=CC=C4)CC5=CN(N=N5)CC6=CC=CC=C6
Applications
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

Solubility
Soluble in DMSO, DMF.

Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.
Melting Point 132°C to 143°C
Color White
Quantity 250mg
Formula Weight 530.63
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine

storageNote1 : -30° to -10°C