Alfa Aesar™ Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4548806

Quantity 5g

 View more versions of this product

Catalog No. AA4548806

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C15H33O6Bi
    Formula Weight 518.4
    MDL Number MFCD11113030
    Chemical Name or Material Tris (1-methoxy-2-methyl-2-propoxy)bismuth

    Specifications

    Formula Weight 518.4
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Health Hazard 1 Warning
    Storage Note 1
    • Packaged under Argon
    Quantity 5g
    Molecular Formula C15H33O6Bi
    MDL Number MFCD11113030
    Color Yellow
    Melting Point 70°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Tris (1-methoxy-2-methyl-2-propoxy)bismuth
    Assay 99.99%, (metals basis)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit