Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%
Click to view available options
Quantity:
250mg
1g
5g
Specifications
Melting Point | 114°C to 116°C |
Boiling Point | 150°C (0.05mmHg) |
Sensitivity | Air and moisture sensitive |
Color | White to Brown |
Quantity | 5g |
Solubility Information | Insoluble in water. |
Formula Weight | 758.79 |
Physical Form | Crystalline |
Percent Purity | 99.9% |
Chemical Name or Material | Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III) |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon