Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99.9% (metals basis)
Click to view available options
Quantity:
1g
5g
Specifications
Boiling Point | 120°C (0.5mmHg)(sublimation) |
Odor | Odorless |
Color | Green |
Quantity | 5g |
Sensitivity | Hygroscopic |
Melting Point | 254°C to 256°C |
Solubility Information | Insoluble in water. |
Formula Weight | 608.72 |
Physical Form | Powder |
Percent Purity | 99.9% |
Show More |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : No
Recommended Storage : Ambient temperatures