Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III), 98%
Specifications
Quantity | 2g |
Sensitivity | Hygroscopic |
Melting Point | 182°C to 183°C |
Formula Weight | 712.32 |
Physical Form | Powder |
Percent Purity | 98% |
Chemical Name or Material | Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 239-563-4
TSCA : No
Recommended Storage : Ambient temperatures