Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III), 98%
Click to view available options
Quantity:
1g
5g
Specifications
Melting Point | 240°C |
Quantity | 1g |
Sensitivity | Hygroscopic |
Boiling Point | 370°C (decomposition) |
Formula Weight | 688.72 |
Physical Form | Powder |
Percent Purity | 98% |
Chemical Name or Material | Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures