Alfa Aesar™ Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), 95+%

Intermediate for scandium oxide ceramics

Manufacturer: Alfa Aesar™ 4278704

Quantity 2g

 View more versions of this product

Catalog No. AA4278704

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15492-49-6
    Molecular Formula C33H57O6Sc
    Formula Weight 594.77
    MDL Number MFCD00169875
    Synonym Scandium(III)-DPM; Sc(TMHD)3
    Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)

    Specifications

    Formula Weight 594.77
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 275°C (dec.)
    Molecular Formula C33H57O6Sc
    CAS 15492-49-6
    Flash Point >110°C (230°F)
    Quantity 2g
    MDL Number MFCD00169875
    Physical Form Powder
    Solubility Information Soluble in ethyl acetate
    Color White
    Sensitivity Hygroscopic
    Synonym Scandium(III)-DPM; Sc(TMHD)3
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 150° to 152°C
    Health Hazard 1 Warning
    Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)scandium (III)
    Assay ≥95%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit