Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals

Catalog Number AC208750050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
50525-27-4
C30H36Cl2N6O6Ru
748.62
MFCD00149670
WHELTKFSBJNBMQ-UHFFFAOYSA-L
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
131664161
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
50525-27-4
C30H36Cl2N6O6Ru
748.62
MFCD00149670
WHELTKFSBJNBMQ-UHFFFAOYSA-L
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
131664161
2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;hydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 748.63
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(2, 2'-bipyridyl)ruthenium(II) chloride, hexahydrate
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit