Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,6-dimethoxyphenyl)phosphine, 97+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
85417-41-0
C24H27O6P
442.448
MFCD00192558
CMLWFCUAXGSMBB-UHFFFAOYSA-N
tris 2,6-dimethoxyphenyl phosphine,tris 2,6-dimethoxyphenyl phosphane,tdmpp,phosphine, tris 2,6-dimethoxyphenyl,tris 2,6-dimethoxyphenly phosphine,pubchem6450,acmc-209q5v,2,6-dmpp,cmlwfcuaxgsmbb-uhfffaoysa,tri 2,6-dimethoxyphenyl phosphine
2734597
tris(2,6-dimethoxyphenyl)phosphane
COC1=C(C(=CC=C1)OC)P(C2=C(C=CC=C2OC)OC)C3=C(C=CC=C3OC)OC
85417-41-0
C24H27O6P
442.448
MFCD00192558
CMLWFCUAXGSMBB-UHFFFAOYSA-N
tris 2,6-dimethoxyphenyl phosphine,tris 2,6-dimethoxyphenyl phosphane,tdmpp,phosphine, tris 2,6-dimethoxyphenyl,tris 2,6-dimethoxyphenly phosphine,pubchem6450,acmc-209q5v,2,6-dmpp,cmlwfcuaxgsmbb-uhfffaoysa,tri 2,6-dimethoxyphenyl phosphine
2734597
tris(2,6-dimethoxyphenyl)phosphane
COC1=C(C(=CC=C1)OC)P(C2=C(C=CC=C2OC)OC)C3=C(C=CC=C3OC)OC
Quantity 5g
Color White
Infrared Spectrum Authentic
Melting Point 151°C to 155°C
Packaging Glass bottle
Assay Percent Range 97+%
Formula Weight 442.45
Physical Form Powder
Percent Purity ≥97.0%
Chemical Name or Material Tris(2, 6-dimethoxyphenyl)phosphine

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.