Learn More
Tris(2-aminoethyl)amine, 97%, Thermo Scientific Chemicals
Description
Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.
Solubility
Miscible with water.
Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
Specifications
Density | 0.978 |
pH | 11.9 |
Boiling Point | 102°C (0.05 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Amine-like |
Refractive Index | 1.4969 |
Linear Formula | N(CH2CH2NH2)3 |
Quantity | 100 g |
UN Number | UN2922 |
Beilstein | 1739626 |
Show More |
Safety and Handling
Hazard Category | H301-H310-H314-H335 |
Hazard Statement | GHS H Statement H301-H310-H314-H318 Toxic if swallowed. Fatal in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P262-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 223-857-4 |
RTECSNumber | KH8587082 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.