Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tris(2-aminoethyl)amine, 97%

Click to view available options
Quantity:
25g
100g
500g
4097-89-6
C6H18N4
146.238
MFCD00008177
MBYLVOKEDDQJDY-UHFFFAOYSA-N
tris 2-aminoethyl amine,tren,taea,2,2',2-triaminotriethylamine,1,2-ethanediamine, n,n-bis 2-aminoethyl,tri 2-aminoethyl amine,tren hp,tris aminoethyl amine,nitrilotris ethylamine,n,n-bis 2-aminoethyl ethane-1,2-diamine
77731
CHEBI:30631
N',N'-bis(2-aminoethyl)ethane-1,2-diamine
C(CN(CCN)CCN)N
4097-89-6
C6H18N4
146.238
MFCD00008177
MBYLVOKEDDQJDY-UHFFFAOYSA-N
tris 2-aminoethyl amine,tren,taea,2,2',2-triaminotriethylamine,1,2-ethanediamine, n,n-bis 2-aminoethyl,tri 2-aminoethyl amine,tren hp,tris aminoethyl amine,nitrilotris ethylamine,n,n-bis 2-aminoethyl ethane-1,2-diamine
77731
CHEBI:30631
N',N'-bis(2-aminoethyl)ethane-1,2-diamine
C(CN(CCN)CCN)N
Applications
Tris(2-aminoethyl)amine is a tetradentate chelating ligand and forms stable complexes with transition metals. It is also used as a carbon dioxide absorbent. Further, it acts as a reagent for cleavage of the fluorenylmethyloxycarbonyl (Fmoc) group in peptide synthesis. It reacts with aryl isocyanates and isothiocyanates to give tris-urea and -thiourea derivatives.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
Refractive Index 1.4969
Sensitivity Air sensitive; Hygroscopic
Density 0.978
Flash Point >110°C (230°F)
Odor Amine-like
Boiling Point 102°C (0.05mmHg)
pH 11.9
Quantity 25g
Linear Formula N(CH2CH2NH2)3
UN Number UN2922
Beilstein 1739626
Solubility Information Miscible with water.
Formula Weight 146.24
Percent Purity 97%
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less

GHS H Statement
H301-H310-H314-H318
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H310-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 223-857-4

RTECSNumber : KH8587082

TSCA : Yes

Recommended Storage : Ambient temperatures