Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-carboxyethyl)phosphine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
51805-45-9
C9H15O6P·HCl
286.65
MFCD00145469
PBVAJRFEEOIAGW-UHFFFAOYSA-N
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
C(CP(CCC(=O)O)CCC(=O)O)C(=O)O.Cl
51805-45-9
C9H15O6P·HCl
286.65
MFCD00145469
PBVAJRFEEOIAGW-UHFFFAOYSA-N
tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
2734570
3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
C(CP(CCC(=O)O)CCC(=O)O)C(=O)O.Cl
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 1g
Assay Percent Range 98%
Beilstein 04, IV, 3441
Merck Index 15, 9222
Solubility Information Solubility in water: soluble.
Formula Weight 286.65
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger