Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris (2-cyanoethyl)phosphine, 95%, Acros Organics

Click to view available options
Quantity:
1g
10g
50g
4023-53-4
C9H12N3P
193.19
CHZAMJVESILJGH-UHFFFAOYSA-N
tris 2-cyanoethyl phosphine,3,3',3-phosphinetriyltripropanenitrile,tris-2-cyanoethyl phosphine,tris-2-cyanoethyl-phosphine,propanenitrile, 3,3',3-phosphinidynetris,acmc-20ajc3,p ch2ch2cn 3,3-bis 2-cyanoethyl phosphanyl propanenitrile,propanenitrile,3',3-phosphinidynetris,tris 2-cyanoethyl phosphine 10g
77639
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
C(CP(CCC#N)CCC#N)C#N
4023-53-4
C9H12N3P
193.19
CHZAMJVESILJGH-UHFFFAOYSA-N
tris 2-cyanoethyl phosphine,3,3',3-phosphinetriyltripropanenitrile,tris-2-cyanoethyl phosphine,tris-2-cyanoethyl-phosphine,propanenitrile, 3,3',3-phosphinidynetris,acmc-20ajc3,p ch2ch2cn 3,3-bis 2-cyanoethyl phosphanyl propanenitrile,propanenitrile,3',3-phosphinidynetris,tris 2-cyanoethyl phosphine 10g
77639
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
C(CP(CCC#N)CCC#N)C#N
Melting Point 97.0°C to 99.0°C
Quantity 10g
Boiling Point 235.0°C (0.9 mmHg)
Packaging Glass bottle
Assay Percent Range 94% min. (Iodimetry)
Solubility Information Solubility in water: slightly soluble
Formula Weight 193.19
Percent Purity 95%
Chemical Name or Material Tris (2-cyanoethyl)phosphine

GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Obtain special instructions before use.
Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Danger

EINECSNumber : 223-687-