Alfa Aesar™ Tris(2-dimethylaminoethyl)amine, 99+%

Manufacturer: Alfa Aesar™ H3185306

Quantity 5g

 View more versions of this product

Catalog No. AAH3185306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 33527-91-2
    Molecular Formula C12H30N4
    Linear Formula [(CH3)2NCH2CH2]3N
    Formula Weight 230.4
    MDL Number MFCD00015607
    UN Number UN2735
    Synonym Me6TREN
    Chemical Name or Material Tris(2-dimethylaminoethyl)amine

    Specifications

    Formula Weight 230.4
    TSCA TSCA
    Linear Formula [(CH3)2NCH2CH2]3N
    UN Number UN2735
    Synonym Me6TREN
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 70° to 71°C (0.5mmHg)
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Molecular Formula C12H30N4
    MDL Number MFCD00015607
    CAS 33527-91-2
    Health Hazard 1 Danger
    Chemical Name or Material Tris(2-dimethylaminoethyl)amine
    Assay ≥99%

    Description

    Applications
    Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.

    Solubility
    Miscible with water.

    Notes
    Hygroscopic. Air sensitive. Incompatible with oxidizing agents, acids.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit