Learn More
Tris(2-dimethylaminoethyl)amine, 98+%, Thermo Scientific Chemicals
Description
Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(2-dimethylaminoethyl)amine is used as an atom transfer radical polymerization (ATRP) ligand for the creation of telechelic polymers. It is a tripodal polyamine ligand and forms complexes with group 1 metals, which is used in alkali-metal-mediated synthetic applications. Further, it is involved in the macrocyclization processes by reacting with tripodal esters in methanol to give tricyclic cryptands.
Solubility
Miscible with water.
Notes
Hygroscopic. Air sensitive. Incompatible with oxidizing agents, acids.
Specifications
Boiling Point | 70°C to 71°C (0.5 mmHg) |
Refractive Index | 1.454 |
Linear Formula | [(CH3)2NCH2CH2]3N |
Quantity | 5 g |
UN Number | UN2735 |
Sensitivity | Air sensitive; Hygroscopic |
Solubility Information | Miscible with water. |
Formula Weight | 230.4 |
Percent Purity | ≥99% |
Chemical Name or Material | Tris(2-dimethylaminoethyl)amine |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.