Alfa Aesar™ Tris(2-methoxyphenyl)phosphine, 98%

Manufacturer: Alfa Aesar™ A1109614

Quantity 25g

 View more versions of this product

Catalog No. AAA1109614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4731-65-1
    Molecular Formula C21H21O3P
    Formula Weight 352.37
    MDL Number MFCD00014892
    Beilstein 2776186
    Synonym Tri(o-anisyl)phosphine; Tris(o-anisyl)phosphine
    Chemical Name or Material Tris (2-methoxyphenyl)phosphine

    Specifications

    Formula Weight 352.37
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C21H21O3P
    Beilstein 2776186
    Health Hazard 1 Warning
    Melting Point 196° to 204°C
    Quantity 25g
    MDL Number MFCD00014892
    CAS 4731-65-1
    Synonym Tri(o-anisyl)phosphine; Tris(o-anisyl)phosphine
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Tris (2-methoxyphenyl)phosphine
    Assay 98%

    Description

    Applications
    Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

    Solubility
    Slightly soluble in water.

    Notes
    Air sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit