Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-methyl-4-diethylaminophenyl)methane, 93+%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4482-70-6
C34H49N3
499.78
OKJSFKIUVDXFMS-UHFFFAOYSA-N
benzenamine, 4,4',4-methylidynetris n,n-diethyl-3-methyl,4,4',4-methylidynetris n,n-diethyl-m-toluidine,tris 2-methyl-4-diethylaminophenyl methane,tris 4-diethylamino-2-methylphenyl methane,4,4',4-methanetriyltris n,n-diethyl-3-methylaniline,4-bis 4-diethylamino-2-methylphenyl methyl-n,n-diethyl-3-methylaniline,4-bis 4-diethylamino-2-methyl-phenyl methyl-n,n-diethyl-3-methyl-aniline
78234
4-[bis[4-(diethylamino)-2-methylphenyl]methyl]-N,N-diethyl-3-methylaniline
CCN(CC)C1=CC(=C(C=C1)C(C2=C(C=C(C=C2)N(CC)CC)C)C3=C(C=C(C=C3)N(CC)CC)C)C
4482-70-6
C34H49N3
499.78
OKJSFKIUVDXFMS-UHFFFAOYSA-N
benzenamine, 4,4',4-methylidynetris n,n-diethyl-3-methyl,4,4',4-methylidynetris n,n-diethyl-m-toluidine,tris 2-methyl-4-diethylaminophenyl methane,tris 4-diethylamino-2-methylphenyl methane,4,4',4-methanetriyltris n,n-diethyl-3-methylaniline,4-bis 4-diethylamino-2-methylphenyl methyl-n,n-diethyl-3-methylaniline,4-bis 4-diethylamino-2-methyl-phenyl methyl-n,n-diethyl-3-methyl-aniline
78234
4-[bis[4-(diethylamino)-2-methylphenyl]methyl]-N,N-diethyl-3-methylaniline
CCN(CC)C1=CC(=C(C=C1)C(C2=C(C=C(C=C2)N(CC)CC)C)C3=C(C=C(C=C3)N(CC)CC)C)C
Melting Point 163°C to 165°C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 93% min. (HPLC)
Linear Formula [(CH3CH2)2NC6H3(CH3)]3CH
Solubility Information Solubility in water: insoluble
Formula Weight 499.78
Percent Purity 93+%
Chemical Name or Material Tris(2-methyl-4-diethylaminophenyl)methane

GHS Signal Word: Warning

EINECSNumber : 224-765-7