Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-thienyl)phosphine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
24171-89-9
C12H9PS3
280.358
MFCD00015471
KUCPTMZJPDVWJL-UHFFFAOYSA-N
tris 2-thienyl phosphine,phosphine, tri-2-thienyl,tri 2-thienyl phosphine,tri thiophen-2-yl phosphine,tri2-thienylphosphine,tris thiophen-2-yl phosphane,tri-2-thienylphosphine,trithienylphosphine,pubchem6427,tri 2-thienyl phosphine #
90384
trithiophen-2-ylphosphane
C1=CSC(=C1)P(C2=CC=CS2)C3=CC=CS3
24171-89-9
C12H9PS3
280.358
MFCD00015471
KUCPTMZJPDVWJL-UHFFFAOYSA-N
tris 2-thienyl phosphine,phosphine, tri-2-thienyl,tri 2-thienyl phosphine,tri thiophen-2-yl phosphine,tri2-thienylphosphine,tris thiophen-2-yl phosphane,tri-2-thienylphosphine,trithienylphosphine,pubchem6427,tri 2-thienyl phosphine #
90384
trithiophen-2-ylphosphane
C1=CSC(=C1)P(C2=CC=CS2)C3=CC=CS3
Quantity 5g
Packaging Glass bottle
Color Green or Peach
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 280.37
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Tris(2-thienyl)phosphine, 98%

AIR SENSITIVE

EINECSNumber : 246-059-8