Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(3-methoxyphenyl)phosphine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
29949-84-6
C21H21O3P
352.37
MFCD00008386
CCXTYQMZVYIQRP-UHFFFAOYSA-N
tris 3-methoxyphenyl phosphine,tris 3-methoxyphenyl phosphane,tris m-methoxyphenyl phosphine,phosphine, tris 3-methoxyphenyl,ksc207e8l,tri m-anisyl phosphine,tris m-anisyl phosphine,phosphine,tris 3-methoxyphenyl,tris-3-methoxy-phenyl-phosphane,phosphine,tris 3-methoxybenzyl
141534
tris(3-methoxyphenyl)phosphane
COC1=CC(=CC=C1)P(C2=CC=CC(=C2)OC)C3=CC=CC(=C3)OC
29949-84-6
C21H21O3P
352.37
MFCD00008386
CCXTYQMZVYIQRP-UHFFFAOYSA-N
tris 3-methoxyphenyl phosphine,tris 3-methoxyphenyl phosphane,tris m-methoxyphenyl phosphine,phosphine, tris 3-methoxyphenyl,ksc207e8l,tri m-anisyl phosphine,tris m-anisyl phosphine,phosphine,tris 3-methoxyphenyl,tris-3-methoxy-phenyl-phosphane,phosphine,tris 3-methoxybenzyl
141534
tris(3-methoxyphenyl)phosphane
COC1=CC(=CC=C1)P(C2=CC=CC(=C2)OC)C3=CC=CC(=C3)OC
Infrared Spectrum Authentic
Quantity 5g
Melting Point 111°C to 114°C
Packaging Glass bottle
Color White
Assay Percent Range 97%
Formula Weight 352.37
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Tris(3-methoxyphenyl)phosphine

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.