Alfa Aesar™ Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate

As a laser dye

Manufacturer: Alfa Aesar™ 4334577

Quantity 0.1g

 View more versions of this product

Catalog No. AA4334577

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14054-87-6
    Molecular Formula C24H12F9O6S3Eu·xH2O
    Formula Weight 815.51 (Anhydrous)
    MDL Number MFCD00041948
    Synonym Europium(III) thenoyltrifluoroacetonate hydrate
    Chemical Name or Material Tris (4,4,4-trifluoro-1- (2-thienyl)-1,3-butanediono)europium (III) hydrate

    Specifications

    Formula Weight 815.51 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 14054-87-6
    Solubility Information Soluble in methanol, toluene
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Yellow
    Melting Point 143° to 144°C
    TSCA TSCA
    Physical Form Powder
    Molecular Formula C24H12F9O6S3Eu·xH2O
    MDL Number MFCD00041948
    Synonym Europium(III) thenoyltrifluoroacetonate hydrate
    Health Hazard 1 Warning
    Quantity 0.1g
    Chemical Name or Material Tris (4,4,4-trifluoro-1- (2-thienyl)-1,3-butanediono)europium (III) hydrate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit