Alfa Aesar™ Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Redox indicator, oxygen sensor

Manufacturer: Alfa Aesar™ 4412304

Quantity 2g

 View more versions of this product

Catalog No. AA4412304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36309-88-3
    Molecular Formula C72H48Cl2N6Ru
    Formula Weight 1169.2
    MDL Number MFCD03095387
    Synonym Tris(bathophenathroline)ruthenium(II) dichloride
    Chemical Name or Material Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride

    Specifications

    Formula Weight 1169.2
    Physical Form Powder
    Molecular Formula C72H48Cl2N6Ru
    Synonym Tris(bathophenathroline)ruthenium(II) dichloride
    MDL Number MFCD03095387
    CAS 36309-88-3
    Quantity 2g
    Chemical Name or Material Tris (4,7-diphenyl-1,10-phenanthroline)ruthenium (II) dichloride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit