Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-iodophenyl)amine 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4181-20-8
C18H12I3N
623.014
MFCD01321198
AQGZDWJFOYXGAA-UHFFFAOYSA-N
tris 4-iodophenyl amine,tris-4-iodophenyl amine,trisiodophenylamine,4-iodo-n,n-bis 4-iodophenyl aniline,tris-4-iodo-phenyl-amine,pubchem19658,tris p-iodophenyl amine,acmc-209jlv,ksc235o2h
9809282
4-iodo-N,N-bis(4-iodophenyl)aniline
C1=CC(=CC=C1N(C2=CC=C(C=C2)I)C3=CC=C(C=C3)I)I
4181-20-8
C18H12I3N
623.014
MFCD01321198
AQGZDWJFOYXGAA-UHFFFAOYSA-N
tris 4-iodophenyl amine,tris-4-iodophenyl amine,trisiodophenylamine,4-iodo-n,n-bis 4-iodophenyl aniline,tris-4-iodo-phenyl-amine,pubchem19658,tris p-iodophenyl amine,acmc-209jlv,ksc235o2h
9809282
4-iodo-N,N-bis(4-iodophenyl)aniline
C1=CC(=CC=C1N(C2=CC=C(C=C2)I)C3=CC=C(C=C3)I)I
Melting Point 186.0°C to 189.0°C
Color Colorless
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula N(C6H4I)3
Formula Weight 623
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Tris(4-iodophenyl)amine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.