Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-trifluoromethylphenyl)phosphine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13406-29-6
C21H12F9P
466.29
MFCD00058883
PXYCJKZSCDFXLR-UHFFFAOYSA-N
tris 4-trifluoromethylphenyl phosphine,tris 4-trifluoromethyl phenyl phosphine,tris 4-trifluoromethyl phenyl phosphane,phosphine, tris 4-trifluoromethyl phenyl,tris p-trifluoromethylphenyl phosphine,phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl,tris p-trifluoromethylphenyl phosphine,min,acmc-20ajbq
139448
tris[4-(trifluoromethyl)phenyl]phosphane
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F
13406-29-6
C21H12F9P
466.29
MFCD00058883
PXYCJKZSCDFXLR-UHFFFAOYSA-N
tris 4-trifluoromethylphenyl phosphine,tris 4-trifluoromethyl phenyl phosphine,tris 4-trifluoromethyl phenyl phosphane,phosphine, tris 4-trifluoromethyl phenyl,tris p-trifluoromethylphenyl phosphine,phosphine, tris .alpha.,.alpha.,.alpha.-trifluoro-p-tolyl,tris p-trifluoromethylphenyl phosphine,min,acmc-20ajbq
139448
tris[4-(trifluoromethyl)phenyl]phosphane
C1=CC(=CC=C1C(F)(F)F)P(C2=CC=C(C=C2)C(F)(F)F)C3=CC=C(C=C3)C(F)(F)F
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Undesignated
Melting Point 72.0°C to 75.0°C
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: insoluble.
Formula Weight 466.29
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tris(4-trifluoromethylphenyl)phosphine
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning