Alfa Aesar™ Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III), 98+%

Manufacturer: Alfa Aesar™ B2175903

Quantity 1g

 View more versions of this product

Catalog No. AAB2175903

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 18323-98-3
    Molecular Formula C30H30DyF21O6
    Formula Weight 1048.04
    MDL Number MFCD00010463
    Synonym Dysprosium-fod
    Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III)

    Specifications

    Formula Weight 1048.04
    MDL Number MFCD00010463
    Synonym Dysprosium-fod
    Melting Point 184° to 186°C
    Quantity 1g
    Molecular Formula C30H30DyF21O6
    CAS 18323-98-3
    Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)dysprosium(III)
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit