Learn More
TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
$106.00 - $437.00

Specifications
Chemical Name or Material | Tris-Acetate-EDTA |
---|---|
Name Note | 1X Solution |
CAS | 77-86-1, 6850-28-8, 139-33-3, 10378-23-1, 7732-18-5 |
Grade | Electrophoresis |
DNase | DNase free |
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
BP243420
|
20L | Poly CUBE |
Each for $437.00
Order in Bulk
Request bulk or custom formats |
|
BP24344
|
4L | Poly CUBE |
Each for $106.00
Order in Bulk
Request bulk or custom formats |
|
Description
TrisAcetateEDTA (TAE) is commonly used as a buffer for nucleic acid electrophoresis.Specifications
Tris-Acetate-EDTA | |
77-86-1, 6850-28-8, 139-33-3, 10378-23-1, 7732-18-5 | |
DNase free | |
Filtered through a 0.2-micron filter. | |
TAE |
1X Solution | |
Electrophoresis | |
Pass Test | |
Protease free |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.