Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS (Base), BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Poly Bottle
Poly Pail
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Pail
Formula Weight 121.14
Quantity 2.5kg
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material TRIS (Base)