Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS (Base), Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
1kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

77-86-1
C4H11NO3
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
CHEBI:9754

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
CAS 77-86-1
Molecular Formula C4H11NO3
Packaging Poly Bottle
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Weight (g/mol) 121.136
PubChem CID 6503
ChEBI CHEBI:9754
Formula Weight 121.14
Quantity 1kg
Chemical Name or Material TRIS (Base)
Grade Ultrapure Bioreagent
Show More Show Less