Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Tris Buffer, 0.3M Solution (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
500mL
Packaging:
Poly Bottle
Description
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 0.3M, provides a convenient stock solution for these applications.
Specifications
Concentration | 0.29 to 0.31M |
Merck Index | 15, 9951 |
DNase | DNase free |
Identification | Pass Test |
Physical Form | Liquid |
Grade | Molecular Biology |
Packaging | Poly Bottle |
Protease | Protease free |
pH | 10.1 to 11.1 |
Color | Undesignated |
Show More |
Safety and Handling

Recommended Storage : RT