Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS-CAPS Buffer, 10X

For blot transfer

$52.80 - $96.90

Specifications

Concentration 10X
Format Liquid
Applications For blot transfer
Product Type Buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ61339K2 View Documents Alfa Aesar™
J61339K2
1L Each for $96.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ61339AP View Documents Alfa Aesar™
J61339AP
500mL Each for $52.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

10X
Liquid
For blot transfer
Buffer
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit