Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Tris(cyclopentadienyl)gadolinium(III), 99.9% (REO)
Click to view available options
Quantity:
1g
5g
Description
- Ampouled under argon
Specifications
Sensitivity | Air and moisture sensitive |
Quantity | 5g |
Packaging | Ampule |
UN Number | UN3181 |
Formula Weight | 352.54 |
Physical Form | Powder |
Percent Purity | 99.9% |
Chemical Name or Material | Tris(cyclopentadienyl)gadolinium(III) |
Assay | (REO) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber : 215-046-9
TSCA : No
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures